Assay Details
| Gene Symbol: LOC4332298 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773722.2 | XP_015629208.1 | 17 - 18 | 1944 | 112 | |
| XM_015773723.2 | XP_015629209.1 | 16 - 17 | 1910 | 112 | ||
| XM_015773724.2 | XP_015629210.1 | 17 - 18 | 1932 | 112 | ||
| XM_015773725.2 | XP_015629211.1 | 17 - 18 | 1944 | 112 | ||
| XM_015773726.2 | XP_015629212.1 | 17 - 18 | 1944 | 112 | ||
| XM_015773728.2 | XP_015629214.1 | 16 - 17 | 1867 | 112 | ||
More Information
Set Membership: |
|

LOC4332298
Genomic Map