Assay Details
| Gene Symbol: LOC4332298 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773722.2 | XP_015629208.1 | 4 - 5 | 396 | 127 | |
| XM_015773723.2 | XP_015629209.1 | 3 - 4 | 362 | 127 | ||
| XM_015773724.2 | XP_015629210.1 | 4 - 5 | 384 | 127 | ||
| XM_015773725.2 | XP_015629211.1 | 4 - 5 | 396 | 127 | ||
| XM_015773726.2 | XP_015629212.1 | 4 - 5 | 396 | 127 | ||
More Information
Set Membership: |
|

LOC4332298
Genomic Map