Assay Details
| Gene Symbol: LOC4332298 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773722.2 | XP_015629208.1 | 8 - 9 | 660 | 65 | |
| XM_015773723.2 | XP_015629209.1 | 7 - 8 | 626 | 65 | ||
| XM_015773724.2 | XP_015629210.1 | 8 - 9 | 648 | 65 | ||
| XM_015773725.2 | XP_015629211.1 | 8 - 9 | 660 | 65 | ||
| XM_015773726.2 | XP_015629212.1 | 8 - 9 | 660 | 65 | ||
| XM_015773728.2 | XP_015629214.1 | 7 - 8 | 583 | 65 | ||
More Information
Set Membership: |
|

LOC4332298
Genomic Map