Assay Details
Gene Symbol: LOC4332314 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775427.2 | XP_015630913.1 | 2 - 3 | 1978 | 134 | |
GenBank mRNA | AK243127.1 | - | 2 - 3 | 1876 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
