Assay Details
| Gene Symbol: LOC4332354 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777334.2 | XP_015632820.1 | 1 - 2 | 151 | 52 | |
| XM_015777336.2 | XP_015632822.1 | 1 - 2 | 151 | 52 | ||
| GenBank mRNA | AK068775.1 | - | 1 - 2 | 93 | 52 | |
| AK104949.1 | - | 1 - 2 | 84 | 52 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015777334.2, XM_015777336.2 transcripts.
Set Membership: |
|

LOC4332354
Genomic Map