Assay Details
| Gene Symbol: LOC4332354 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777334.2 | XP_015632820.1 | 4 - 5 | 445 | 117 | |
| XM_015777336.2 | XP_015632822.1 | 4 - 5 | 445 | 117 | ||
| GenBank mRNA | AK068775.1 | - | 4 - 5 | 387 | 117 | |
| AK104949.1 | - | 4 - 5 | 378 | 117 | ||
More Information
Set Membership: |
|

LOC4332354
Genomic Map