Assay Details
| Gene Symbol: LOC4332471 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775995.2 | XP_015631481.1 | 10 - 11 | 2145 | 64 | |
| GenBank mRNA | AK060735.1 | - | 8 - 9 | 1298 | 64 | |
| AK068146.1 | - | 10 - 11 | 2153 | 64 | ||
| AK099067.1 | - | 10 - 11 | 2123 | 64 | ||
More Information
Set Membership: |
|

LOC4332471
Genomic Map