Assay Details
| Gene Symbol: LOC4332471 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775995.2 | XP_015631481.1 | 11 - 12 | 2210 | 88 | |
| GenBank mRNA | AK060735.1 | - | 9 - 10 | 1363 | 88 | |
| AK065726.1 | - | 6 - 7 | 923 | 88 | ||
| AK068146.1 | - | 11 - 12 | 2218 | 88 | ||
| AK099067.1 | - | 11 - 12 | 2188 | 88 | ||
More Information
Set Membership: |
|

LOC4332471
Genomic Map