Assay Details
| Gene Symbol: LOC4332471 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775995.2 | XP_015631481.1 | 2 - 3 | 669 | 105 | |
| GenBank mRNA | AK068146.1 | - | 2 - 3 | 677 | 105 | |
| AK099067.1 | - | 2 - 3 | 647 | 105 | ||
More Information
Set Membership: |
|

LOC4332471
Genomic Map