Assay Details
| Gene Symbol: LOC4332471 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775995.2 | XP_015631481.1 | 5 - 6 | 1146 | 150 | |
| GenBank mRNA | AK060735.1 | - | 3 - 4 | 299 | 150 | |
| AK068146.1 | - | 5 - 6 | 1154 | 150 | ||
| AK099067.1 | - | 5 - 6 | 1124 | 150 | ||
More Information
Set Membership: |
|

LOC4332471
Genomic Map