Assay Details
| Gene Symbol: LOC4332474 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775012.2 | XP_015630498.1 | 6 - 7 | 669 | 99 | |
| GenBank mRNA | AK061841.1 | - | 6 - 7 | 644 | 99 | |
| BT130287.1 | - | 6 - 7 | 645 | 99 | ||
| GQ848050.1 | - | 6 - 7 | 636 | 99 | ||
More Information
Set Membership: |
|

LOC4332474
Genomic Map