Assay Details
| Gene Symbol: LOC4333161 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774943.2 | XP_015630429.1 | 1 - 2 | 218 | 75 | |
| GenBank mRNA | AK069123.1 | - | 1 - 2 | 225 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015774943.2 transcripts.
Set Membership: |
|

LOC4333161
Genomic Map