Assay Details
| Gene Symbol: LOC4333316 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773024.2 | XP_015628510.1 | 1 | 1544 | 126 | |
| GenBank mRNA | AK061586.1 | - | 1 | 1471 | 126 | |
| AK072995.1 | - | 1 | 1549 | 126 | ||
More Information
Set Membership: |
|

LOC4333316
Genomic Map