Assay Details
| Gene Symbol: LOC4333368 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773148.2 | 1 - 2 | 1108 | 57 | ||
| GenBank mRNA | AK069017.1 | - | 1 - 2 | 972 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4333368| Gene Symbol: LOC4333368 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773148.2 | 1 - 2 | 1108 | 57 | ||
| GenBank mRNA | AK069017.1 | - | 1 - 2 | 972 | 57 | |
Set Membership: |
|