Assay Details
| Gene Symbol: LOC4333522 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773471.2 | XP_015628957.1 | 1 - 2 | 296 | 57 | |
| GenBank mRNA | AK073129.1 | - | 1 - 2 | 174 | 57 | |
| AK073903.1 | - | 1 - 2 | 297 | 57 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015773471.2 transcripts.
Set Membership: |
|

LOC4333522
Genomic Map