Assay Details
| Gene Symbol: LOC4333554 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774903.2 | XP_015630389.1 | 1 - 2 | 375 | 122 | |
| GenBank mRNA | AK099445.1 | - | 1 - 2 | 313 | 122 | |
| AK108010.1 | - | 1 - 2 | 342 | 122 | ||
More Information
Set Membership: |
|

LOC4333554
Genomic Map