Assay Details
| Gene Symbol: LOC4333554 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774903.2 | XP_015630389.1 | 4 - 5 | 872 | 113 | |
| GenBank mRNA | AK099445.1 | - | 4 - 5 | 810 | 113 | |
More Information
Set Membership: |
|

LOC4333554
Genomic Map