Assay Details
| Gene Symbol: LOC107280215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772789.2 | XP_015628275.1 | 2 - 3 | 917 | 69 | |
| XM_015772790.2 | XP_015628276.1 | 3 - 4 | 650 | 69 | ||
| XM_015772791.2 | XP_015628277.1 | 3 - 4 | 573 | 69 | ||
| XM_015772794.2 | XP_015628280.1 | 2 - 3 | 917 | 69 | ||
| XM_015772795.2 | XP_015628281.1 | 2 - 3 | 917 | 69 | ||
More Information
Set Membership: |
|

LOC107280215
Genomic Map