Assay Details
| Gene Symbol: LOC107280215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772789.2 | XP_015628275.1 | 5 - 6 | 1101 | 80 | |
| XM_015772790.2 | XP_015628276.1 | 6 - 7 | 834 | 80 | ||
| XM_015772791.2 | XP_015628277.1 | 6 - 7 | 757 | 80 | ||
More Information
Set Membership: |
|

LOC107280215
Genomic Map