Assay Details
| Gene Symbol: LOC4334232 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776201.2 | XP_015631687.1 | 2 - 3 | 354 | 71 | |
| GenBank mRNA | AK066137.1 | - | 2 - 3 | 317 | 71 | |
| AK071361.1 | - | 2 - 3 | 308 | 71 | ||
| AK103989.1 | - | 2 - 3 | 316 | 71 | ||
More Information
Set Membership: |
|

LOC4334232
Genomic Map