Assay Details
| Gene Symbol: LOC4334321 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774602.2 | XP_015630088.1 | 1 | 1179 | 99 | |
| GenBank mRNA | AK069119.1 | - | 1 | 1148 | 99 | |
More Information
Set Membership: |
|

LOC4334321
Genomic Map