Assay Details
| Gene Symbol: LOC4334593 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773786.2 | XP_015629272.1 | 1 | 554 | 151 | |
| GenBank mRNA | AK119690.1 | - | 1 | 549 | 151 | |
| AY305864.1 | - | 1 | 537 | 151 | ||
More Information
Set Membership: |
|

LOC4334593
Genomic Map