Assay Details
| Gene Symbol: LOC4334609 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777479.2 | XP_015632965.1 | 3 - 4 | 348 | 77 | |
| XM_015777480.2 | XP_015632966.1 | 3 - 4 | 336 | 77 | ||
| GenBank mRNA | AK060050.1 | - | 3 - 4 | 303 | 77 | |
More Information
Set Membership: |
|

LOC4334609
Genomic Map