Assay Details
| Gene Symbol: LOC4334803 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776075.2 | XP_015631561.1 | 1 - 2 | 606 | 97 | |
| XM_015776076.2 | XP_015631562.1 | 1 - 2 | 610 | 97 | ||
| GenBank mRNA | AK070236.1 | - | 1 - 2 | 329 | 97 | |
| AK071441.1 | - | 1 - 2 | 389 | 97 | ||
More Information
Set Membership: |
|

LOC4334803
Genomic Map