Assay Details
| Gene Symbol: LOC4334803 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776075.2 | XP_015631561.1 | 2 - 3 | 638 | 120 | |
| XM_015776076.2 | XP_015631562.1 | 2 - 3 | 642 | 120 | ||
| GenBank mRNA | AK070236.1 | - | 2 - 3 | 361 | 120 | |
| AK071441.1 | - | 2 - 3 | 421 | 120 | ||
More Information
Set Membership: |
|

LOC4334803
Genomic Map