Assay Details
Gene Symbol: LOC4334969 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015781152.2 | XP_015636638.1 | 3 - 4 | 1129 | 80 | |
GenBank mRNA | AJ698084.1 | - | 3 - 4 | 646 | 80 | |
AK061898.1 | - | 3 - 4 | 643 | 80 | ||
AK065035.1 | - | 3 - 4 | 888 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
