Assay Details
| Gene Symbol: LOC4335004 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778054.2 | XP_015633540.1 | 1 - 2 | 3555 | 66 | |
| GenBank mRNA | AK112081.1 | - | 1 - 2 | 3000 | 66 | |
| AK121779.1 | - | 1 - 2 | 3289 | 66 | ||
More Information
Set Membership: |
|

LOC4335004
Genomic Map