Assay Details
| Gene Symbol: LOC4335112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780494.2 | XP_015635980.1 | 3 - 4 | 387 | 144 | |
| GenBank mRNA | AK064939.1 | - | 3 - 4 | 348 | 144 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015780494.2 transcripts.
Set Membership: |
|

LOC4335112
Genomic Map