Assay Details
| Gene Symbol: LOC4335112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780494.2 | XP_015635980.1 | 7 - 8 | 2498 | 84 | |
| XM_015780496.2 | XP_015635982.1 | 6 - 7 | 2404 | 84 | ||
| GenBank mRNA | AK064939.1 | - | 7 - 8 | 2459 | 84 | |
More Information
Set Membership: |
|

LOC4335112
Genomic Map