Assay Details
| Gene Symbol: LOC4335638 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781690.1 | XP_015637176.1 | 2 - 3 | 1339 | 78 | |
| GenBank mRNA | AK108218.1 | - | 1 - 2 | 963 | 78 | |
More Information
Set Membership: |
|

LOC4335638
Genomic Map