Assay Details
| Gene Symbol: LOC4335640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780363.2 | XP_015635849.1 | 3 - 4 | 742 | 111 | |
| GenBank mRNA | AK071912.1 | - | 3 - 4 | 619 | 111 | |
| AK101795.1 | - | 3 - 4 | 735 | 111 | ||
| AK104588.1 | - | 2 - 3 | 288 | 111 | ||
| AK104732.1 | - | 2 - 3 | 288 | 111 | ||
More Information
Set Membership: |
|

LOC4335640
Genomic Map