Assay Details
| Gene Symbol: LOC4335640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780363.2 | XP_015635849.1 | 5 - 6 | 1192 | 105 | |
| GenBank mRNA | AK071912.1 | - | 5 - 6 | 1069 | 105 | |
| AK101795.1 | - | 5 - 6 | 1185 | 105 | ||
| AK104588.1 | - | 4 - 5 | 738 | 105 | ||
| AK104732.1 | - | 4 - 5 | 738 | 105 | ||
More Information
Set Membership: |
|

LOC4335640
Genomic Map