Assay Details
| Gene Symbol: LOC4336044 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780502.1 | XP_015635988.1 | 2 - 3 | 272 | 95 | |
| GenBank mRNA | AK071685.1 | - | 2 - 3 | 192 | 95 | |
| AK103922.1 | - | 2 - 3 | 196 | 95 | ||
| AK103932.1 | - | 2 - 3 | 196 | 95 | ||
| AK103994.1 | - | 2 - 3 | 196 | 95 | ||
| AK104690.1 | - | 2 - 3 | 196 | 95 | ||
| AK119220.1 | - | 2 - 3 | 191 | 95 | ||
| AK119522.1 | - | 2 - 3 | 191 | 95 | ||
| AK241494.1 | - | 197 | 95 | |||
| AK287771.1 | - | 197 | 95 | |||
More Information
Set Membership: |
|

LOC4336044
Genomic Map