Assay Details
| Gene Symbol: LOC9268615 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778519.2 | XP_015634005.1 | 2 - 3 | 370 | 132 | |
| GenBank mRNA | AK065850.1 | - | 2 - 3 | 378 | 132 | |
More Information
Set Membership: |
|

LOC9268615
Genomic Map