Assay Details
| Gene Symbol: LOC9268615 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778519.2 | XP_015634005.1 | 4 - 5 | 550 | 64 | |
| XM_015778520.2 | XP_015634006.1 | 4 - 5 | 493 | 64 | ||
| GenBank mRNA | AK065850.1 | - | 4 - 5 | 558 | 64 | |
More Information
Set Membership: |
|

LOC9268615
Genomic Map