Assay Details
| Gene Symbol: LOC9268615 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778519.2 | XP_015634005.1 | 8 - 9 | 986 | 78 | |
| XM_015778520.2 | XP_015634006.1 | 8 - 9 | 929 | 78 | ||
| GenBank mRNA | AK065850.1 | - | 8 - 9 | 994 | 78 | |
More Information
Set Membership: |
|

LOC9268615
Genomic Map