Assay Details
| Gene Symbol: LOC4336057 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780820.2 | XP_015636306.1 | 6 - 7 | 781 | 92 | |
| GenBank mRNA | AK243669.1 | - | 5 - 6 | 1472 | 92 | |
More Information
Set Membership: |
|

LOC4336057
Genomic Map