Assay Details
| Gene Symbol: LOC4336085 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778068.2 | XP_015633554.1 | 1 - 2 | 636 | 63 | |
| XM_015778069.2 | XP_015633555.1 | 1 - 2 | 636 | 63 | ||
| XM_015778070.2 | XP_015633556.1 | 1 - 2 | 636 | 63 | ||
| XM_015778071.2 | XP_015633557.1 | 1 - 2 | 647 | 63 | ||
| XM_015778073.2 | XP_015633559.1 | 1 - 2 | 649 | 63 | ||
More Information
Set Membership: |
|

LOC4336085
Genomic Map