Assay Details
| Gene Symbol: LOC4336129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778632.2 | XP_015634118.1 | 6 - 7 | 1454 | 103 | |
| GenBank mRNA | AK070336.1 | - | 3 - 4 | 430 | 103 | |
| AK070339.1 | - | 6 - 7 | 1455 | 103 | ||
More Information
Set Membership: |
|

LOC4336129
Genomic Map