Assay Details
| Gene Symbol: LOC4336257 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780844.1 | XP_015636330.1 | 20 - 21 | 2619 | 87 | |
| XM_015780845.2 | XP_015636331.1 | 18 - 19 | 2840 | 87 | ||
| XM_015780846.1 | XP_015636332.1 | 19 - 20 | 2565 | 87 | ||
| XM_015780847.1 | XP_015636333.1 | 20 - 21 | 2538 | 87 | ||
| GenBank mRNA | AK072226.1 | - | 2 - 3 | 161 | 87 | |
More Information
Set Membership: |
|

LOC4336257
Genomic Map