Assay Details
| Gene Symbol: LOC4336386 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781305.2 | XP_015636791.1 | 1 - 2 | 1680 | 88 | |
| GenBank mRNA | AK067204.1 | - | 1 - 2 | 1560 | 88 | |
More Information
Set Membership: |
|

LOC4336386
Genomic Map