Assay Details
| Gene Symbol: LOC9271254 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778996.2 | XP_015634482.1 | 1 - 2 | 184 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC9271254| Gene Symbol: LOC9271254 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778996.2 | XP_015634482.1 | 1 - 2 | 184 | 92 | |
Set Membership: |
|