Assay Details
| Gene Symbol: LOC4336656 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779725.1 | XP_015635211.1 | 1 - 2 | 173 | 90 | |
| GenBank mRNA | AK107862.1 | - | 1 - 2 | 175 | 90 | |
More Information
Set Membership: |
|

LOC4336656
Genomic Map