Assay Details
| Gene Symbol: LOC4336656 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779725.1 | XP_015635211.1 | 2 - 3 | 455 | 79 | |
| GenBank mRNA | AK107862.1 | - | 2 - 3 | 457 | 79 | |
More Information
Set Membership: |
|

LOC4336656
Genomic Map