Assay Details
| Gene Symbol: LOC4337061 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779130.2 | 3 - 4 | 936 | 76 | ||
| GenBank mRNA | AK062930.1 | - | 2 - 3 | 235 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4337061| Gene Symbol: LOC4337061 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779130.2 | 3 - 4 | 936 | 76 | ||
| GenBank mRNA | AK062930.1 | - | 2 - 3 | 235 | 76 | |
Set Membership: |
|