Assay Details
| Gene Symbol: LOC4337676 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783856.2 | XP_015639342.1 | 11 - 12 | 1206 | 105 | |
| GenBank mRNA | AK066314.1 | - | 11 - 12 | 1069 | 105 | |
| AK071923.1 | - | 1232 | 105 | |||
More Information
Set Membership: |
|

LOC4337676
Genomic Map