Assay Details
| Gene Symbol: LOC4337676 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783856.2 | XP_015639342.1 | 5 - 6 | 703 | 110 | |
| GenBank mRNA | AK066314.1 | - | 5 - 6 | 566 | 110 | |
| AK071923.1 | - | 729 | 110 | |||
More Information
Set Membership: |
|

LOC4337676
Genomic Map