Assay Details
| Gene Symbol: LOC4337696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785175.2 | XP_015640661.1 | 7 - 8 | 843 | 139 | |
| XM_015785176.2 | XP_015640662.1 | 7 - 8 | 833 | 139 | ||
| GenBank mRNA | AJ508916.2 | - | 6 - 7 | 954 | 139 | |
More Information
Set Membership: |
|

LOC4337696
Genomic Map