Assay Details
| Gene Symbol: LOC4338670 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782258.2 | XP_015637744.1 | 2 - 3 | 350 | 84 | |
| GenBank mRNA | AK059190.1 | - | 2 - 3 | 288 | 84 | |
More Information
Set Membership: |
|

LOC4338670
Genomic Map