Assay Details
| Gene Symbol: LOC4338736 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785064.2 | XP_015640550.1 | 2 - 3 | 517 | 76 | |
| GenBank mRNA | AK070043.1 | - | 2 - 3 | 505 | 76 | |
| AK104199.1 | - | 2 - 3 | 504 | 76 | ||
| AK104366.1 | - | 2 - 3 | 504 | 76 | ||
More Information
Set Membership: |
|

LOC4338736
Genomic Map